Assay Details
| Gene Symbol: NUFIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK349801.1 | - | 1 | 417 | 86 | |
| AK352353.1 | - | 1 | 403 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NUFIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK349801.1 | - | 1 | 417 | 86 | |
| AK352353.1 | - | 1 | 403 | 86 | ||
Set Membership: |
|