Assay Details
| Gene Symbol: KRTAP11-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358920.4 | XP_003358968.1 | 823 | 110 | ||
| GenBank mRNA | AK349903.1 | - | 1 | 822 | 110 | |
More Information
Set Membership: |
|

KRTAP11-1
Genomic Map