Assay Details
| Gene Symbol: CCN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091372.1 | XP_020947031.1 | 490 | 59 | ||
| XM_021091378.1 | XP_020947037.1 | 357 | 59 | |||
| XM_021091382.1 | XP_020947041.1 | 501 | 59 | |||
| XM_021091387.1 | XP_020947046.1 | 499 | 59 | |||
| GenBank mRNA | AK349957.1 | - | 1 - 2 | 134 | 59 | |
More Information
Set Membership: |
|

CCN6
Genomic Map