Assay Details
| Gene Symbol: CCN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091382.1 | XP_020947041.1 | 1043 | 128 | ||
| XM_021091387.1 | XP_020947046.1 | 1041 | 128 | |||
| GenBank mRNA | AK349957.1 | - | 4 - 5 | 774 | 128 | |
More Information
Set Membership: |
|

CCN6
Genomic Map