Assay Details
| Gene Symbol: CST6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353767.2 | XP_003353815.1 | 508 | 93 | ||
| GenBank mRNA | AK349968.1 | - | 2 - 3 | 377 | 93 | |
| AK397234.1 | - | 2 - 3 | 381 | 93 | ||
| AY610298.1 | - | 2 - 3 | 375 | 93 | ||
More Information
Set Membership: |
|

CST6
Genomic Map