Assay Details
| Gene Symbol: LOXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021074114.1 | XP_020929773.1 | 2289 | 56 | ||
| XM_021074115.1 | XP_020929774.1 | 2307 | 56 | |||
| GenBank mRNA | AK350047.1 | - | 10 - 11 | 2128 | 56 | |
More Information
Set Membership: |
|

LOXL2
Genomic Map