Assay Details
Gene Symbol: BEND7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021064846.1 | XP_020920505.1 | 1140 | 57 | ||
XM_021064847.1 | XP_020920506.1 | 1284 | 57 | |||
XM_021064848.1 | XP_020920507.1 | 1140 | 57 | |||
XM_021064849.1 | XP_020920508.1 | 1140 | 57 | |||
XM_021064850.1 | XP_020920509.1 | 1140 | 57 | |||
XM_021064851.1 | XP_020920510.1 | 1140 | 57 | |||
XM_021064853.1 | XP_020920512.1 | 1140 | 57 | |||
XM_021064854.1 | XP_020920513.1 | 1261 | 57 | |||
XM_021064855.1 | XP_020920514.1 | 1200 | 57 | |||
XM_021064856.1 | XP_020920515.1 | 1140 | 57 | |||
XM_021064857.1 | XP_020920516.1 | 1199 | 57 | |||
XM_021064858.1 | XP_020920517.1 | 1201 | 57 | |||
GenBank mRNA | AK350058.1 | - | 4 - 5 | 725 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
