Assay Details
| Gene Symbol: AP1M2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013987439.2 | XP_013842893.2 | 1007 | 56 | ||
| XM_021083790.1 | XP_020939449.1 | 1097 | 56 | |||
| GenBank mRNA | AK350095.1 | - | 8 - 9 | 931 | 56 | |
More Information
Set Membership: |
|

AP1M2
Genomic Map