Assay Details
| Gene Symbol: CSRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481768.4 | XP_003481816.2 | 2418 | 67 | ||
| XM_021092813.1 | XP_020948472.1 | 497 | 67 | |||
| XM_021092814.1 | XP_020948473.1 | 2422 | 67 | |||
| GenBank mRNA | AK350342.1 | - | 4 - 5 | 971 | 67 | |
| AY610113.1 | - | 4 - 5 | 492 | 67 | ||
More Information
Set Membership: |
|

CSRP2
Genomic Map