Assay Details
| Gene Symbol: TNFRSF13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021068044.1 | XP_020923703.1 | 518 | 95 | ||
| GenBank mRNA | AK350344.1 | - | 3 - 4 | 456 | 95 | |
| AK397710.1 | - | 3 - 4 | 450 | 95 | ||
More Information
Set Membership: |
|

TNFRSF13B
Genomic Map