Assay Details
| Gene Symbol: MSC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125595.4 | XP_003125643.1 | 765 | 86 | ||
| GenBank mRNA | AK350490.1 | - | 1 - 2 | 768 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MSC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125595.4 | XP_003125643.1 | 765 | 86 | ||
| GenBank mRNA | AK350490.1 | - | 1 - 2 | 768 | 86 | |
Set Membership: |
|