Assay Details
| Gene Symbol: ZBTB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021062868.1 | XP_020918527.1 | 1673 | 56 | ||
| XM_021062869.1 | XP_020918528.1 | 1596 | 56 | |||
| XM_021062870.1 | XP_020918529.1 | 1473 | 56 | |||
| GenBank mRNA | AK350495.1 | - | 3 - 4 | 1169 | 56 | |
More Information
Set Membership: |
|

ZBTB16
Genomic Map