Assay Details
| Gene Symbol: LONRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077473.1 | XP_020933132.1 | 3449 | 128 | ||
| GenBank mRNA | AK350664.1 | - | 1 | 918 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LONRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077473.1 | XP_020933132.1 | 3449 | 128 | ||
| GenBank mRNA | AK350664.1 | - | 1 | 918 | 128 | |
Set Membership: |
|