Assay Details
Gene Symbol: LOC100515166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131436.3 | XP_003131484.1 | 1325 | 96 | ||
GenBank mRNA | AK350696.1 | - | 6 - 7 | 1304 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
