Assay Details
| Gene Symbol: MGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097378.1 | XP_020953037.1 | 7318 | 64 | ||
| GenBank mRNA | AK350700.1 | - | 2 - 3 | 992 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097378.1 | XP_020953037.1 | 7318 | 64 | ||
| GenBank mRNA | AK350700.1 | - | 2 - 3 | 992 | 64 | |
Set Membership: |
|