Assay Details
| Gene Symbol: CCDC162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090803.1 | XP_020946462.1 | 6069 | 85 | ||
| XM_021090805.1 | XP_020946464.1 | 6064 | 85 | |||
| XM_021090807.1 | XP_020946466.1 | 6419 | 85 | |||
| XM_021090809.1 | XP_020946468.1 | 6109 | 85 | |||
| XM_021090812.1 | XP_020946471.1 | 6459 | 85 | |||
| XM_021090819.1 | XP_020946478.1 | 6060 | 85 | |||
| XM_021090827.1 | XP_020946486.1 | 6069 | 85 | |||
| XM_021090832.1 | XP_020946491.1 | 5855 | 85 | |||
| XM_021090834.1 | XP_020946493.1 | 5931 | 85 | |||
| XM_021090835.1 | XP_020946494.1 | 6069 | 85 | |||
| XM_021090836.1 | XP_020946495.1 | 4883 | 85 | |||
| GenBank mRNA | AK350846.1 | - | 6 - 7 | 727 | 85 | |
More Information
Set Membership: |
|

CCDC162
Genomic Map