Assay Details
| Gene Symbol: CCDC162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090803.1 | XP_020946462.1 | 6700 | 101 | ||
| XM_021090805.1 | XP_020946464.1 | 6695 | 101 | |||
| XM_021090807.1 | XP_020946466.1 | 7050 | 101 | |||
| XM_021090809.1 | XP_020946468.1 | 6740 | 101 | |||
| XM_021090812.1 | XP_020946471.1 | 7090 | 101 | |||
| XM_021090816.1 | XP_020946475.1 | 6697 | 101 | |||
| XM_021090819.1 | XP_020946478.1 | 6691 | 101 | |||
| XM_021090827.1 | XP_020946486.1 | 6685 | 101 | |||
| XM_021090829.1 | XP_020946488.1 | 6682 | 101 | |||
| XM_021090832.1 | XP_020946491.1 | 6486 | 101 | |||
| XM_021090834.1 | XP_020946493.1 | 6562 | 101 | |||
| XM_021090835.1 | XP_020946494.1 | 6547 | 101 | |||
| XM_021090836.1 | XP_020946495.1 | 5514 | 101 | |||
| GenBank mRNA | AK350846.1 | - | 9 - 10 | 1133 | 101 | |
More Information
Set Membership: |
|

CCDC162
Genomic Map