Assay Details
| Gene Symbol: NR2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083574.1 | XP_020939233.1 | 939 | 55 | ||
| GenBank mRNA | AK350867.1 | - | 1 - 2 | 437 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NR2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083574.1 | XP_020939233.1 | 939 | 55 | ||
| GenBank mRNA | AK350867.1 | - | 1 - 2 | 437 | 55 | |
Set Membership: |
|