Assay Details
| Gene Symbol: NR2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083574.1 | XP_020939233.1 | 1036 | 58 | ||
| GenBank mRNA | AK350867.1 | - | 2 - 3 | 534 | 58 | |
More Information
Set Membership: |
|

NR2F6
Genomic Map