Assay Details
| Gene Symbol: IQCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013984621.2 | XP_013840075.2 | 1971 | 59 | ||
| GenBank mRNA | AK350917.1 | - | 4 - 5 | 778 | 59 | |
| AK393902.1 | - | 4 - 5 | 893 | 59 | ||
| AK399303.1 | - | 3 - 4 | 632 | 59 | ||
More Information
Set Membership: |
|

IQCA1
Genomic Map