Assay Details
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131817.5 | XP_003131865.2 | 692 | 56 | ||
XM_021067531.1 | XP_020923190.1 | 692 | 56 | |||
GenBank mRNA | AK350926.1 | - | 1 - 2 | 47 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
