Assay Details
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131817.5 | XP_003131865.2 | 779 | 98 | ||
GenBank mRNA | AK350926.1 | - | 2 - 3 | 134 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
