Assay Details
| Gene Symbol: CDHR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013979834.2 | XP_013835288.2 | 1145 | 77 | ||
| GenBank mRNA | AK350950.1 | - | 5 - 6 | 743 | 77 | |
More Information
Set Membership: |
|

CDHR3
Genomic Map