Assay Details
| Gene Symbol: ZMYM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065423.1 | XP_020921082.1 | 493 | 64 | ||
| XM_021065424.1 | XP_020921083.1 | 227 | 64 | |||
| XM_021065425.1 | XP_020921084.1 | 195 | 64 | |||
| GenBank mRNA | AK351084.1 | - | 2 - 3 | 173 | 64 | |
| AK400808.1 | - | 2 - 3 | 153 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021065425.1 transcripts.
Set Membership: |
|

ZMYM5
Genomic Map