Assay Details
| Gene Symbol: AIF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353688.4 | XP_003353736.2 | 195 | 58 | ||
| GenBank mRNA | AK351089.1 | - | 2 - 3 | 189 | 58 | |
| AK392602.1 | - | 2 - 3 | 189 | 58 | ||
More Information
Set Membership: |
|

AIF1L
Genomic Map