Assay Details
| Gene Symbol: OVOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122528.4 | XP_003122576.1 | 2370 | 90 | ||
| XM_021082825.1 | XP_020938484.1 | 2155 | 90 | |||
| GenBank mRNA | AK351093.1 | - | 1 | 612 | 90 | |
More Information
Set Membership: |
|

OVOL1
Genomic Map