Assay Details
| Gene Symbol: NT5C3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066994.1 | XP_020922653.1 | 860 | 67 | ||
| XM_021066995.1 | XP_020922654.1 | 485 | 67 | |||
| GenBank mRNA | AK351119.1 | - | 5 - 6 | 655 | 67 | |
| AK351352.1 | - | 5 - 6 | 647 | 67 | ||
More Information
Set Membership: |
|

NT5C3B
Genomic Map