Assay Details
| Gene Symbol: RPP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927117.5 | XP_001927152.1 | 3981 | 59 | ||
| GenBank mRNA | AK351259.1 | - | 1 | 1227 | 59 | |
| AK390222.1 | - | 1 | 1253 | 59 | ||
More Information
Set Membership: |
|

RPP25
Genomic Map