Assay Details
| Gene Symbol: WHAMM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927315.5 | XP_001927350.1 | 919 | 98 | ||
| GenBank mRNA | AK351300.1 | - | 2 - 3 | 888 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: WHAMM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927315.5 | XP_001927350.1 | 919 | 98 | ||
| GenBank mRNA | AK351300.1 | - | 2 - 3 | 888 | 98 | |
Set Membership: |
|