Assay Details
| Gene Symbol: HIVEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659167.3 | XP_005659224.1 | 9840 | 111 | ||
| XM_021072375.1 | XP_020928034.1 | 35305 | 111 | |||
| XM_021072379.1 | XP_020928038.1 | 35385 | 111 | |||
| XM_021072385.1 | XP_020928044.1 | 35131 | 111 | |||
| XM_021072389.1 | XP_020928048.1 | 35230 | 111 | |||
| XM_021072392.1 | XP_020928051.1 | 9915 | 111 | |||
| XM_021072400.1 | XP_020928059.1 | 9707 | 111 | |||
| XM_021072405.1 | XP_020928064.1 | 9865 | 111 | |||
| XM_021072410.1 | XP_020928069.1 | 9691 | 111 | |||
| XM_021072419.1 | XP_020928078.1 | 35464 | 111 | |||
| XM_021072421.1 | XP_020928080.1 | 9790 | 111 | |||
| XM_021072427.1 | XP_020928086.1 | 9514 | 111 | |||
| XM_021072434.1 | XP_020928093.1 | 9616 | 111 | |||
| XM_021072441.1 | XP_020928100.1 | 35214 | 111 | |||
| GenBank mRNA | AK351843.1 | - | 1 | 1017 | 111 | |
More Information
Set Membership: |
|

HIVEP2
Genomic Map