Assay Details
| Gene Symbol: HNRNPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095510.1 | XP_020951169.1 | 844 | 65 | ||
| XM_021095511.1 | XP_020951170.1 | 836 | 65 | |||
| XM_021095512.1 | XP_020951171.1 | 832 | 65 | |||
| XM_021095513.1 | XP_020951172.1 | 1251 | 65 | |||
| XM_021095514.1 | XP_020951173.1 | 1112 | 65 | |||
| XM_021095515.1 | XP_020951174.1 | 675 | 65 | |||
| XM_021095516.1 | XP_020951175.1 | 670 | 65 | |||
| GenBank mRNA | AK351947.1 | - | 5 - 6 | 591 | 65 | |
| AK393356.1 | - | 6 - 7 | 761 | 65 | ||
More Information
Set Membership: |
|

HNRNPR
Genomic Map