Assay Details
| Gene Symbol: MED31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358216.4 | XP_003358264.1 | 311 | 73 | ||
| GenBank mRNA | AK351969.1 | - | 1 - 2 | 121 | 73 | |
| AK391837.1 | - | 1 - 2 | 81 | 73 | ||
More Information
Set Membership: |
|

MED31
Genomic Map