Assay Details
| Gene Symbol: LOC100525452 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084681.1 | XP_020940340.1 | 3142 | 116 | ||
| GenBank mRNA | AK352117.1 | - | 1 - 2 | 150 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021084681.1 transcripts.
Set Membership: |
|

LOC100525452
Genomic Map