Assay Details
| Gene Symbol: GOSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131316.6 | XP_003131364.1 | 196 | 68 | ||
| XM_021066828.1 | XP_020922487.1 | 196 | 68 | |||
| GenBank mRNA | AK352256.1 | - | 2 - 3 | 156 | 68 | |
More Information
Set Membership: |
|

GOSR2
Genomic Map