Assay Details
| Gene Symbol: FYB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096577.1 | XP_020952236.1 | 1328 | 96 | ||
| XM_021096578.1 | XP_020952237.1 | 1325 | 96 | |||
| XM_021096579.1 | XP_020952238.1 | 1328 | 96 | |||
| XM_021096580.1 | XP_020952239.1 | 1328 | 96 | |||
| GenBank mRNA | AK352268.1 | - | 6 - 7 | 1209 | 96 | |
| AK396675.1 | - | 6 - 7 | 1257 | 96 | ||
More Information
Set Membership: |
|

FYB2
Genomic Map