Assay Details
| Gene Symbol: CYB5RL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096598.1 | XP_020952257.1 | 573 | 93 | ||
| XM_021096599.1 | XP_020952258.1 | 565 | 93 | |||
| XM_021096600.1 | XP_020952259.1 | 668 | 93 | |||
| XM_021096601.1 | XP_020952260.1 | 740 | 93 | |||
| XM_021096602.1 | XP_020952261.1 | 574 | 93 | |||
| XM_021096603.1 | XP_020952262.1 | 573 | 93 | |||
| GenBank mRNA | AK352402.1 | - | 2 - 3 | 548 | 93 | |
More Information
Set Membership: |
|

CYB5RL
Genomic Map