Assay Details
| Gene Symbol: RBM12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125566.4 | XP_003125614.1 | 3674 | 76 | ||
| XM_005662972.3 | XP_005663029.1 | 3652 | 76 | |||
| XM_013996610.2 | XP_013852064.1 | 3486 | 76 | |||
| XM_013996611.2 | XP_013852065.1 | 3888 | 76 | |||
| XM_021089110.1 | XP_020944769.1 | 4031 | 76 | |||
| XM_021089111.1 | XP_020944770.1 | 4427 | 76 | |||
| XM_021089112.1 | XP_020944771.1 | 3602 | 76 | |||
| GenBank mRNA | AK389178.1 | - | 1 | 2421 | 76 | |
More Information
Set Membership: |
|

RBM12B
Genomic Map