Assay Details
| Gene Symbol: ATRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005669525.3 | XP_005669582.3 | 763 | 69 | ||
| GenBank mRNA | AK389232.1 | - | 2 - 3 | 299 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ATRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005669525.3 | XP_005669582.3 | 763 | 69 | ||
| GenBank mRNA | AK389232.1 | - | 2 - 3 | 299 | 69 | |
Set Membership: |
|