Assay Details
| Gene Symbol: EHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087658.1 | XP_020943317.1 | 3875 | 124 | ||
| GenBank mRNA | AK389234.1 | - | 1 | 1923 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087658.1 | XP_020943317.1 | 3875 | 124 | ||
| GenBank mRNA | AK389234.1 | - | 1 | 1923 | 124 | |
Set Membership: |
|