Assay Details
| Gene Symbol: ZFP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083007.1 | XP_020938666.1 | 1922 | 106 | ||
| XM_021083008.1 | XP_020938667.1 | 1238 | 106 | |||
| GenBank mRNA | AK389239.1 | - | 2 - 3 | 144 | 106 | |
| AK395415.1 | - | 10 - 11 | 970 | 106 | ||
More Information
Set Membership: |
|

ZFP91
Genomic Map