Assay Details
| Gene Symbol: FZD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005667210.3 | XP_005667267.2 | 2686 | 123 | ||
| GenBank mRNA | AK231613.1 | - | 1 | 125 | 123 | |
| AK235104.1 | - | 1 | 287 | 123 | ||
| AK393695.1 | - | 1 | 521 | 123 | ||
More Information
Set Membership: |
|

FZD4
Genomic Map