Assay Details
| Gene Symbol: POLR1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013991968.2 | XP_013847422.2 | 716 | 111 | ||
| GenBank mRNA | AK231695.1 | - | 1 - 2 | 78 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013991968.2 transcripts.
Set Membership: |
|

POLR1G
Genomic Map