Assay Details
| Gene Symbol: RPL27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482496.4 | XP_003482544.2 | 1358 | 93 | ||
| GenBank mRNA | AK231703.1 | - | 3 - 4 | 166 | 93 | |
| AK392163.1 | - | 3 - 4 | 167 | 93 | ||
| AK398167.1 | - | 3 - 4 | 1140 | 93 | ||
| AK399793.1 | - | 3 - 4 | 169 | 93 | ||
More Information
Set Membership: |
|

RPL27A
Genomic Map