Assay Details
| Gene Symbol: RPL27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482496.4 | XP_003482544.2 | 1533 | 91 | ||
| GenBank mRNA | AK231703.1 | - | 4 - 5 | 341 | 91 | |
| AK392163.1 | - | 4 - 5 | 342 | 91 | ||
| AK398167.1 | - | 4 - 5 | 1315 | 91 | ||
| AK399793.1 | - | 4 - 5 | 344 | 91 | ||
More Information
Set Membership: |
|

RPL27A
Genomic Map