Assay Details
| Gene Symbol: CYB5D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131870.4 | XP_003131918.1 | 554 | 70 | ||
| XM_005669123.3 | XP_005669180.1 | 559 | 70 | |||
| XM_021067697.1 | XP_020923356.1 | 557 | 70 | |||
| GenBank mRNA | AK231717.1 | - | 1 - 2 | 540 | 70 | |
More Information
Set Membership: |
|

CYB5D2
Genomic Map