Assay Details
| Gene Symbol: NKX3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483391.4 | XP_003483439.1 | 404 | 56 | ||
| GenBank mRNA | AK231780.1 | - | 1 - 2 | 401 | 56 | |
| AK396395.1 | - | 1 - 2 | 401 | 56 | ||
More Information
Set Membership: |
|

NKX3-1
Genomic Map