Assay Details
| Gene Symbol: LDLRAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005665262.3 | XP_005665319.1 | 1160 | 74 | ||
| XM_005665263.3 | XP_005665320.1 | 1396 | 74 | |||
| XM_013999207.2 | XP_013854661.1 | 1173 | 74 | |||
| XM_013999208.2 | XP_013854662.1 | 1368 | 74 | |||
| XM_013999210.2 | XP_013854664.1 | 1030 | 74 | |||
| XM_013999211.2 | XP_013854665.1 | 1281 | 74 | |||
| XM_013999212.2 | XP_013854666.1 | 1410 | 74 | |||
| XM_021096016.1 | XP_020951675.1 | 868 | 74 | |||
| XM_021096017.1 | XP_020951676.1 | 864 | 74 | |||
| GenBank mRNA | AK231795.1 | - | 6 - 7 | 949 | 74 | |
More Information
Set Membership: |
|

LDLRAD4
Genomic Map