Assay Details
| Gene Symbol: RO60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK231823.1 | - | 6 - 7 | 811 | 98 | |
| AK238342.1 | - | 7 - 8 | 1487 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RO60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK231823.1 | - | 6 - 7 | 811 | 98 | |
| AK238342.1 | - | 7 - 8 | 1487 | 98 | ||
Set Membership: |
|