Assay Details
| Gene Symbol: DHRS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095382.1 | XP_020951041.1 | 1391 | 71 | ||
| GenBank mRNA | AK231831.1 | - | 2 - 3 | 712 | 71 | |
| AK232772.1 | - | 2 - 3 | 715 | 71 | ||
| AK233160.1 | - | 2 - 3 | 719 | 71 | ||
| AK394166.1 | - | 2 - 3 | 722 | 71 | ||
| AK396410.1 | - | 2 - 3 | 753 | 71 | ||
| AK397494.1 | - | 2 - 3 | 749 | 71 | ||
More Information
Set Membership: |
|

DHRS3
Genomic Map