Assay Details
| Gene Symbol: TXNL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK231952.1 | - | 1 | 1423 | 94 | |
| AK352036.1 | - | 1 | 1370 | 94 | ||
| AK400827.1 | - | 2 | 2174 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TXNL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK231952.1 | - | 1 | 1423 | 94 | |
| AK352036.1 | - | 1 | 1370 | 94 | ||
| AK400827.1 | - | 2 | 2174 | 94 | ||
Set Membership: |
|